You Searched For: Vacuum Hand Pumps
4  results were found
SearchResultCount:"4"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: BÃœRKLE
Description: The vacuum hand pump can be operated with one hand to create a vacuum or pressure, independent of power supply.
Supplier: Thermo Fisher Scientific
Description: With these sturdy pumps, a partial vacuum of 86 mbar can be reached very quickly and easily. The vacuum can be controlled with just the index finger. ...
Supplier: KARTELL
Description: Vacuum pumps have built in vacuum release valve allows for quick and easy release of the vacuum without disconnecting the pump.
Catalog Number: (SART16673)
Supplier: Sartorius
Description: The hand operated vacuum pump made of PVC is used as practical vacuum source for outside of a laboratory.
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 4 of 4
no targeter for Bottom