Magnetic stirrer, lab disc white
Supplier: IKA
442-0737EA 366 EUR
442-0737 IKAA3765000
Magnetic stirrer, lab disc white
Stirrers Magnetic Stirrers
Ultra-flat magnetic stirrers with magnetic coil technology. Top plate and casing are made of chemically resistant materials. Wear-free drive with no moving parts.
  • Reverses direction of rotation every 30 seconds to ensure optimum mixing (selectable)
  • Can be used at 5 to 40 °C (80% relative humidity), in an incubator or cold room
Certifikace: IP protection class according to DIN EN 60529: IP 65