Orbital shaker, mini, SSM1
Supplier: Cole Parmer
stuart®
444-0117EA 1430 EUR
444-0117
Orbital shaker, mini, SSM1
Shakers Orbital Shakers
The compact SSM1 provides a smooth uniform circular motion, which is ideal for samples from 0,5 to 5 ml in multiwell plates, dishes and Petri dishes. For larger vessels, an optional accessory cradle system is available that can accommodate a variety of vessels including flasks, bottles or beakers via four rubber securing bars. It will hold up to 4×250 ml or 2×500 ml or 1×1000 ml Erlenmeyer flasks or bottles. Once set on the digital display, the shaking speed is effectively maintained even over long periods of time.
  • Built-in digital timer
  • Variable speed control to 300 min⁻¹
  • Digital timer which can be set can be set in hours, minutes, seconds or for continuous operation
  • Can be used at 4 to 40 °C (80% relative humidity), in an incubator or cold room
  • BioCote® antimicrobial protection
Certifikace: IP Protection Class according to DIN EN 60529: IP 31
Informace pro objednávání: Supplied with non-slip mat for up to four multi-well plates or diagnostic cards.