You Searched For: Shakers
231  results were found
SearchResultCount:"231"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Shaking incubators with a small footprint. These compact units feature a stackable design and expanded temperature range which makes the ILS 4 and ILS...
Catalog Number: (444-0270)
Supplier: VWR Collection
Description: Microplate shaker with microprocessor control which holds two or four microplates, deep well plates or two microtube racks. Variable speed across a wi...
UOM: 1 * 1 items
Waste Reducing
Catalog Number: (444-7106)
Supplier: VWR Collection
Description: A compact, powerful mini shaker with microprocessor control that is ideal for a wide variety of shaking applications. The unit has optimal speed range...
UOM: 1 * 1 items
Waste Reducing
Supplier: HAVER & BOECKER
Description: The EML 200 series offers different types of units. All models are suitable for 50 to 203 mm Ø sieves, a maximum sample/batch load of 3 kg, and have a...
New Product
Supplier: HAVER & BOECKER
Description: These electromagnetically-driven test sieve shakers are designed for sieve analysis with optimum three-dimensional sieving action. The self-re-adjusti...
New Product
Catalog Number: (444-2906)
Supplier: VWR Collection
Description: Digital shaker with microprocessor control which is ideal for a range mixing applications, e.g. cell cultures, solubility studies and extraction proce...
UOM: 1 * 1 items
Waste Reducing
Catalog Number: (444-0762)
Supplier: VWR Collection
Description: The incubating rocking platform shaker combines smooth rocking motion and incubation in one compact bench top unit. The spill resistant design channel...
UOM: 1 * 1 items
Catalog Number: (444-0272)
Supplier: VWR Collection
Description: Incubating microplate shaker with microprocessor control which holds up to four microplates or two microtube racks. The speed can be adjusted across a...
UOM: 1 * 1 items
Waste Reducing
Supplier: VWR Collection
Description: Ergonomic mini vortex mixers with features to suit different applications, these compact units provide optimum comfort and minimal stress.
Waste Reducing
Supplier: VWR Collection
Description: Designed to hold multiple test tubes, test tube racks, vials or cylinders. Vortexing action is created by securing the top of the vessel in place whil...
Catalog Number: (444-1372)
Supplier: VWR Collection
Description: This vortex mixer combines state of the art electronics with design and comfort. Start the vortexing action by simply pressing down on the holder or p...
UOM: 1 * 1 items
Supplier: VWR Collection
Description: The VWR® Standard microplate vortex mixer is a variable speed analogue unit with high speed and small orbit for effectively mixing microplates. The VW...
Catalog Number: (444-0004)
Supplier: VWR Collection
Description: Vortex mixer suitable for single, small containers up to 30 mm in diameter, for example, test tubes, centrifuge tubes and microtubes. The upper casing...
UOM: 1 * 1 items
Catalog Number: (444-2900)
Supplier: VWR Collection
Description: General purpose shaker with microprocessor control which is ideal for basic mixing applications, blotting techniques, staining and destaining. Variabl...
UOM: 1 * 1 items
Waste Reducing
Supplier: VWR Collection
Description: General purpose shaker with microprocessor control which is ideal for basic mixing applications, bacterial suspensions, staining and destaining. Varia...
Catalog Number: (444-1343)
Supplier: VWR Collection
Description: The VWR® rotating mixer provides gentle to vigorous mixing of samples in up to 50 ml tubes.
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 231
no targeter for Bottom