You Searched For: Fluid Samplers
59  results were found
SearchResultCount:"59"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: Avantor Fluid Handling
Description: WFI Samplers are an easy and convenient solution for obtaining perfect fluid samples from a WFI water drop.
Catalog Number: (231-0500)
Supplier: VWR Collection
Description: White PTFE ladle.
UOM: 1 * 1 items
Catalog Number: (9815-05)
Supplier: Avantor
Description: Cycle-Tap sampler is a device for sampling small quantities of solvent.
UOM: 1 * 1 items
Supplier: BÃœRKLE
Description: ViscoSampler with powerful suction for viscous media. The media is drawn up and discharged using the suction flask.
Supplier: Spectrum Laboratories
Description: Disposable tubes for liquid and solid samples with one hand. The sample to be transfered is inserted with harvester and discharged using the plunger.
Supplier: NASCO FORT ATKINSON
Description: The Sludge Judge line of collection tools are dependable and long-lasting.
Supplier: BÃœRKLE
Description: ProfiSampler, the vacuum-operated sample collector for robust industrial use.
Supplier: BÃœRKLE
Description: PP.Ideal para la toma de muestras de piscinas, depósitos, bocas de alcantarillas, aguas superficiales. Adecuado para pozos profundos y estrechos. El m...
Supplier: BÃœRKLE
Description: Material: V4A stainless steel (1.4404). Sampler for thin to viscous liquids.
Catalog Number: (300-1041)
Supplier: BÃœRKLE
Description: The collection lance consists of an inner tube and an outer tube. The suction position can be varied steplessly with the outer tube within a height of...
UOM: 1 * 1 items
Catalog Number: (BURK5365-6000)
Supplier: BÃœRKLE
Description: Precise liquid samples can be taken with this immersion cylinder. When the target has been reached, open the cover and pour out the sample.
UOM: 1 * 1 items
Supplier: BÃœRKLE
Description: Made of ultra-pure and chemically inert PTFE/FEP or PP for contamination-free sampling. Fits all vats and barrels with openings of at least 32 mm Ø.
Supplier: BÃœRKLE
Description: Complete handy set combining the LiquiSampler and the ViscoSampler with practical accessories.
Catalog Number: (300-1042)
Supplier: BÃœRKLE
Description: Hand operated vacuum pump sampler with a 2,5 metre long PVC hose, the stainless steel weight will sink the end of the hose to the required sampling de...
UOM: 1 * 1 items
Catalog Number: (300-0665)
Supplier: BÃœRKLE
Description: Mini immersion cylinder for sampling liquids in narrow places where access is difficult.
UOM: 1 * 1 items
Supplier: BÃœRKLE
Description: The sample liquid only comes into contact with the suction hose and the original sample bottle. Cross-contamination is greatly reduced. Immediately af...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 59
no targeter for Bottom