You Searched For: Air Samplers
18  results were found
SearchResultCount:"18"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: SAS Super Pinocchio CR is a compact microbial impaction sampler designed to test the quality of compressed air and gases used in cleanrooms. A compres...
Supplier: VWR Collection
Description: The SAS Super IAQ provides the same efficiency as recorded by sampling tests conducted by Centre for Applied Microbiology & Research (CAMR-UK).
Supplier: VWR Collection
Description: SAS Super Pinocchio II is a microbial impaction sampler to test the quality of compressed air and gases used in cleanrooms. A compressed air source is...
Supplier: VWR Collection
Description: When environmental contamination is very low, and the number of samples to be performed is high, operational duration can be an important factor in th...
Supplier: VWR Collection
Description: The SAS instrument control unit is placed outside and connected to the aspirating head using a simple electrical connection. All possible typical cont...
Supplier: VWR Collection
Description: Environmental monitoring in cleanrooms is important and a critical part of the production chain for a manufacturing company.
Supplier: VWR Collection
Description: SAS Super ISO 100 and SAS Super ISO 180 are differentiated by their airflow; this choice depends on the application, and allows continuous or sequenti...
Catalog Number: (710-1973)
Supplier: Bertin Instruments
Description: Coriolis® μ is a portable biological air sampler for indoor or outdoor air bio-contamination monitoring. The equipment concentrates airborne particles...
UOM: 1 * 1 items
Supplier: Bertin Instruments
Description: Portable dry air sampler for bio contamination control.
Supplier: Restek
Description: Sample inlet, 152 mm, Siltek® treated stainless steel
Supplier: Restek
Description: These passive air samplers provide accurate integrated sampling without a sampling pump.
Supplier: Restek
Description: These small canisters are designed for controlled sampling, as an alternative to tube and pump samplers.
Supplier: Restek
Description: These grab air samplers are suitable for use with canisters.
Supplier: Restek
Description: These air samplers provide accurate integrated sampling via small fittings that easily connect to miniature canisters.
Supplier: Restek
Description: This unique grab sampler minimises connections and leaks and houses a critical orifice in the tee fitting.
Supplier: Restek
Description: PUF plugs fits to glass holders as an alternative to pre-cleaned packed cartridges.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 18
no targeter for Bottom