Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
Refrigerators
https://si.vwr.com/stibo/smallweb/std.lang.all/97/28/18989728.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"51"
  List View Easy View (new)
Sort by:
Supplier:  LIEBHERR
Description:   Commercial refrigerators with forced air cooling; the ventilation stops when door is opened.
Supplier:  LIEBHERR
Description:   Table top refrigerators for general use.
Supplier:  LIEBHERR
Description:   These commercial refrigerators feature a fan for forced air cooling with temperature adjustable from –2 to +15 °C or +1 to +15 °C.
Supplier:  LIEBHERR
Description:   Laboratory refrigerators with ventilated cooling at +5 °C ±3 °C, Cofrac certified.
Supplier:  LIEBHERR
Description:   These combined fridge freezers with biofresh and nofrost offers more interior capacity for food storage, economy and energy efficiency.
UOM:  1 * 1 items
Supplier:  LIEBHERR
Description:   Laboratory refrigerators with ventilated cooling 5 °C, ±3 °C.
Catalog Number: (LIEBBCDV1003)
Supplier:  LIEBHERR
Description:   These commercial refrigerators have large glass doors, optimum lighting, and height-adjustable shelves. Manufactured with the highest-quality material...
UOM:  1 * 1 items
Supplier:  LIEBHERR
Description:   This MedilLne laboratory refrigerator line includes TC 02 command, which consists of an electronic circuit control, located behind the front panel or...
UOM:  1 * 1 items
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8  of 51
  1  2  3  4  5  6  7  Next 
A new look and a renewed commitment
Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.
Learn More Start Shopping