MediaJet automated Petri dish filler
Supplier: Integra Biosciences
MediaJet
710-1525EA 0 EUR
710-1525 INTG113000 710-1524 INTG103006 INTG103005
MediaJet automated Petri dish filler
Petri Dish Fillers
The Mediajet Vario is the perfect filler for a wide range of Petri dish sizes; it can be adapted to fill Petri dishes of 60 or 90 mm diameters in minutes. The Mediajet Vario is a hands-free system with the highest possible capacity in a limited space. In conjunction with the MediaClave media preparator, ±540 standard Petri dishes can be prepared and filled in approximately 2,5 hours.
  • Petri dishes are mechanically guided throughout the process by a reliable 'walk-away' system in order to avoid damaging them
  • Via the software, the height of the Petri dish can be very easily programmed (between 12 and 25 mm), setting Mediajet apart from other Petri dish brands
  • The 'agar spread' function allows less medium to be used per dish, thereby reducing costs
Functions are controlled and programmed via a clear and simple graphic display (available in six languages).
This simple and adaptable device can be fitted with a carousel for 540 standard Petri dishes.
Fitted with a UV lamp along the entire length of the rotor.
Compatible with an Inkjet printer to automatically label Petri dishes.
The 'Tubefiller' option enables you to switch from Petri dish filling to automatic tube filling in less than a minute.