VWR® VENTI-Line®, Ovens with Forced Convection
Supplier: VWR
VWR®
466-3519EA 0 EUR
466-3519
VWR® VENTI-Line®, Ovens with Forced Convection
Ovens Standard Ovens
Drying ovens with forced air circulation, which are ideal for routine drying and heating applications when short drying times are required. Ovens feature adjustable ventilation by means of manually adjustable ventilation slide and 50 mm Ø exhaust duct at the rear of the unit.
  • Microprocessor controller
  • Integrated timer can be set for continuous mode or timer operation: 0 to 999 min or 0 to 99,9 h
  • Over-temperature protection with visual alarm (Class 3.1 safety device)
  • Temperature setting in increments of 1 °C
Delivery information: Supplied with two chrome plated shelves.