Mixer mills, MM 200 and MM 400
Supplier: Retsch
412-2176EA 7020 EUR
412-2176
Mixer mills, MM 200 and MM 400
Mills
These simple to use laboratory mills have been developed for grinding and homogenising small amounts of sample. They are suitable for size reduction of hard and brittle samples as well as for soft, elastic and fibrous materials, disruption of biological cells and DNA/RNA recovery. The MM 400 can also be used for wet and cryogenic grinding. The grinding time is very short and the sample is hardly warmed at all. Application areas include mineralogy, chemical and environmental research as well as pharmaceutical sample analysis and biotechnology. Cell disruption can be achieved in reaction vials using specially designed adapters. The mills can also mix and homogenise powders and suspensions in less than a minute.
  • Two grinding stations can process two samples simultaneously
  • Digital setting of grinding time and vibration intensity ensures reproducible results
  • Nine parameter combinations can be stored
  • Screw top grinding jars (up to 50 ml, MM 400 only) allow safe wet and cryogenic grinding without loss of material
  • Self-locking clamping device and self-centering for grinding jars makes use simple and safe (MM 400)
* Depending on feed material and instrument configuration/settings
Certifikace: IP Protection Class according to DIN EN 60529: IP 30
Informace pro objednávání: Grinding jars and grinding balls are available in a wide selection of sizes and materials, these need to be ordered separately.
Upozornení: * Depending on feed material and instrument configuration/settings.