You Searched For: Light Meters
5  results were found
SearchResultCount:"5"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (662-0940)
Supplier: VWR Collection
Description: Dual-displays show lux or foot-candles, or percent differential from a reference point, and the selected light source (daylight, tungsten, fluorescent...
UOM: 1 * 1 items
Catalog Number: (TEST0563.1545)
Supplier: Testo
Description: Lux meter testo 545 can measure the illuminance of all common light sources quickly, easily and precisely to ensure productivity, performance and heal...
UOM: 1 * 1 items
Supplier: UVP ULTRA VIOLET PRODUCTS LTD
Description: The J-Series UV meters are photovoltaic devices for taking repeatable readings of UV light sources in either the 365 nm or 254 nm UV range.
Catalog Number: (UVPA97-0015-02)
Supplier: UVP ULTRA VIOLET PRODUCTS LTD
Description: This digital UVX Radiometer is used with one of the three interchangeable sensors (ordered separately) for measuring 254, 365 and 302 nm UV wavelength...
UOM: 1 * 1 items
Catalog Number: (620-3304)
Supplier: Testo
Description: This luxmeter permits measurements of light intensity. It provides a sensor adjusted to the spectral sensitivity of the eye. A backlit display and an ...
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 5 of 5
no targeter for Bottom