You Searched For: Level Sensors
39  results were found
SearchResultCount:"39"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: Avantor
Description: These level indicators provide the digital readout of the approximate volume of solvent remaining in the CYCLE-TAINER system.
Supplier: S.C.A.T.
Description: Monitor two filling levels at once, perfect integration into lab furniture with the Built-In Signalbox ant the Signalbox T1 and T4.
Supplier: S.C.A.T.
Description: Automatically divert waste liquids into the reserve canister.
Supplier: Bohlender
Description: Insert with screw-in thread for barrels or screw cap for canisters. With integrated level indicator to monitor the liquid level during filling as well...
Supplier: Bohlender
Description: Display unit to monitor connected fill-level sensors in b.safe Waste Caps and b.safe Waste Caps.
Supplier: S.C.A.T.
Description: Measuring filling levels continuously. Monitoring non-transparent canisters on a permanent basis is no problem with the continuous level control unit.
Supplier: Bohlender
Description: Display unit to control the b.safe Splitter, which is available as an option in combination with connected fill-level sensors in b.safe Waste Caps and...
Catalog Number: (BOHLR640-05)
Supplier: Bohlender
Description: Optional device for fill level or empty level control in containers and canisters made of non-conductive materials.
UOM: 1 * 1 items
Catalog Number: (MSGA13299)
Supplier: asecos
Description: This level indicator is suitable for all standard 200 l barrels and non-corrosive liquids.
UOM: 1 * 1 items
Supplier: Bohlender
Description: Display unit to monitor connected fill-level sensors in b.safe Waste Caps and b.safe System Waste Caps.
Supplier: Bohlender
Description: The b.safe Mini-display is the optimal basic solution for the local control of single HPLC units.
Supplier: Bohlender
Description: The b.safe Mini-Display is the optimal basic solution for the local control of single HPLC units.
Supplier: S.C.A.T.
Description: These level sensors provide dependable protection from overflow when filling barrels. The float positions itself securely at the opening by its own we...
Supplier: S.C.A.T.
Description: Monitor two filling levels at once. Perfect integration into lab furniture with the Built-In signalbox and switch amplifier.
Catalog Number: (BOHLR660-01)
Supplier: Bohlender
Description: Enclosed housing for installation at the rear side of a safety cabinet. Unit provides safe signal exchange between explosion-hazardous and explosion-p...
UOM: 1 * 1 items
Catalog Number: (BOHLR653-01)
Supplier: Bohlender
Description: Alarm box with aluminium case for an installation in front plates of lab furniture. Display unit to control the b.safe Splitter, which is available as...
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 39
no targeter for Bottom