Order Entry
Ice cubers, self contained, ACM and EC series
 :  SCOTSMAN
New Product
undefined
Ice cubers, self contained, ACM and EC series
Supplier:  SCOTSMAN
New Product
ACM ice cubers
  •  
    Seq
    Water consumption
    Ice capacity (l)
    Storage capacity
    W×D×H
    Weight
  • Ice maker, ACM 106
    Ice maker, ACM 106
    474-1409
    ACM106
    04
    136 l/h
    50 L/24 h
    23 kg
    530×600×970 mm
    52 kg
    1 * 1 items
    1 * 1 items
    Accessories
    Specifications
    Environmentally Preferable
  • Ice maker, ACM 107
    Ice maker, ACM 107
    SCOMACM107
    ACM107
    05
    136 l/h
    23 kg
    531×600×930 mm
    54 kg
    1 * 1 items
    1 * 1 items - 2 346,00 €
    2 346,00 €
    1 * 1 items
       2 346,00 €
    New Product
    Accessories
    Specifications
    Environmentally Preferable
    New Product
  • Ice maker, ACM 126
    Ice maker, ACM 126
    474-1410
    ACM126
    06
    190 l/h
    71 L/24 h
    39 kg
    680×600×1050 mm
    75 kg
    1 * 1 items
    1 * 1 items
    Accessories
    Specifications
    Environmentally Preferable
  • Ice maker, ACM 176
    Ice maker, ACM 176
    474-1411
    ACM176
    07
    150 l/h
    84 L/24 h
    48 kg
    680×600×1120 mm
    80 kg
    1 * 1 items
    1 * 1 items
    Accessories
    Specifications
    Environmentally Preferable
  • Ice maker, ACM 46
    Ice maker, ACM 46
    474-1412
    ACM46
    01
    77 l/h
    24 L/24 h
    9 kg
    386×600×765 mm
    35 kg
    1 * 1 items
    1 * 1 items - 1 720,00 €
    1 720,00 €
    1 * 1 items
       1 720,00 €
    Accessories
    Specifications
    Environmentally Preferable
  • Ice maker, ACM 56
    Ice maker, ACM 56
    474-1413
    ACM56
    02
    89 l/h
    31 L/24 h
    12,5 kg
    386×600×830 mm
    36 kg
    1 * 1 items
    1 * 1 items
    Accessories
    Specifications
    Environmentally Preferable
  • Ice maker, ACM 86
    Ice maker, ACM 86
    474-1414
    ACM86
    03
    144 l/h
    38 L/24 h
    19 kg
    530×600×915 mm
    44 kg
    1 * 1 items
    1 * 1 items
    Accessories
    Specifications
    Environmentally Preferable
EC ice cubers
  •  
    Seq
    Water consumption
    Ice capacity (l)
    Storage capacity
    W×D×H
    Weight
  • Ice maker with built in waste drain pump, EC 46
    Ice maker with built in waste drain pump, EC 46
    SCOMEC46
    EC46
    08
    77 l/h
    24 L/24 h
    9 kg
    386×600×765 mm
    35 kg
    1 * 1 items
    1 * 1 items
    Accessories
    Specifications
    Environmentally Preferable
  • Ice maker with built in waste drain pump, EC 86
    Ice maker with built in waste drain pump, EC 86
    SCOMEC86
    EC86
    09
    144 l/h
    38 L/24 h
    19 kg
    530×600×915 mm
    44 kg
    1 * 1 items
    1 * 1 items
    Accessories
    Specifications
    Environmentally Preferable
  • Ice maker with built in waste drain pump, EC 106
    Ice maker with built in waste drain pump, EC 106
    SCOMEC106
    EC106
    10
    136 l/h
    50 L/24 h
    23 kg
    530×600×970 mm
    52 kg
    1 * 1 items
    1 * 1 items
    Accessories
    Specifications
    Environmentally Preferable
Water filter system
  •  
    Seq
    Description
    Pk
  • 050
    Water filter system, HF20-S
    1
    1 * 1 items
    1 * 1 items - 278,00 €
    278,00 €