You Searched For: Ice Makers
9  results were found
SearchResultCount:"9"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: SCOTSMAN
Description: ACM ice cubers:
The ACM series self contained ice cube makers are air cooled, electronically controlled and manufactured from 304 stainles...
Supplier: SCOTSMAN
Description: The EF self contained ice flakers are air-cooled and manufactured from 304 stainless steel.
Supplier: SCOTSMAN
Description: The AF self contained ice flakers are air-cooled and manufactured from 304 stainless steel.
Supplier: SCOTSMAN
Description: Modular ice flakers is designed to produce hygienic, clear, pure ice ideal for medical and laboratory applications. It requires both a water and elect...
Catalog Number: (474-0036)
Supplier: SCOTSMAN
Description: The TC180 ice cube and water dispenser is a fully automatic air cooled dispensing station that dispenses ice cubelets or water or both simultaneously ...
UOM: 1 * 1 items
Catalog Number: (474-0022)
Supplier: SCOTSMAN
Description: The ACM25, Easy Ice, self contained ice cube maker is a manual water fill, air cooled unit that is manufactured from injection moulded plastic. It pro...
UOM: 1 * 1 items
Catalog Number: (SCOMAFE424)
Supplier: SCOTSMAN
Description: AFE424 undercounter ice flakers with three-piece cabinets are constructed of durable rust-free rotocats plastic.
UOM: 1 * 1 items
Catalog Number: (BELAF388740000)
Supplier: Bel-Art Products, a Part of SP
Description: This compact device can be attached to a liquid COâ‚‚ cylinder (not included) equipped with a dip tube to produce a ready-to-use 250 to 350 g block of d...
UOM: 1 * 1 items
Catalog Number: (HOSHFM-170AKE)
Supplier: HOSHIZAKI
Description: The FM-170AKE (-N) is a modular ice machine that, depending on the model, produces flake ice or nugget ice.
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 9 of 9
no targeter for Bottom