Order Entry
Laboratory blenders, Masticator
undefined
Laboratory blenders, Masticator
Supplier:  IUL Instruments SA
Specifications
  • Catalog No:
  • 710-0104
  • 710-0101
  • 710-0103
  • 710-0100
  • 710-0102
  • 710-0105
  • Model:
  • Classic/Basic
  • Silver
  • Classic/Basic
  • Classic/Basic
  • Classic/Basic
  • Silver
  • Motor speed:
  • 1500 min⁻¹
  • 500…1500 min⁻¹
  • 1500 min⁻¹
  • 1500 min⁻¹
  • 1500 min⁻¹
  • 500…1500 min⁻¹
  • Reduction:
  • 1:6
  • 1:6
  • 1:6
  • 1:6
  • 1:6
  • 1:6
  • Strokes:
  • 8,5/s
  • 5/6/7/8/s
  • 8,5/s
  • 8,5/s
  • 8,5/s
  • 5/6/7/8/s
  • Timer:
  • 15 - 90 seconds; digital setting, continuous mode
  • 5/30/60/90/120/150/180/210/240 s; keyboard digital setting, continuous mode
  • 15 - 90 seconds; digital setting, continuous mode
  • 15 - 90 seconds; digital setting, continuous mode
  • 15 - 90 seconds; digital setting, continuous mode
  • 5/30/60/90/120/150/180/210/240 s; keyboard digital setting, continuous mode
  • Start/stop:
  • door activated
  • door activated
  • door activated
  • door activated
  • door activated
  • door activated
  • Power:
  • 270 W
  • 270 W
  • 270 W
  • 270 W
  • 270 W
  • 270 W
  • Motor:
  • Induction
  • Induction
  • Induction
  • Induction
  • Induction
  • Induction
  • W×D×H:
  • 240×400×260 mm
  • 240×400×300 mm
  • 240×400×260 mm
  • 240×400×260 mm
  • 240×400×260 mm
  • 240×400×300 mm
  • Weight:
  • 17 kg
  • 20 kg
  • 17 kg
  • 17 kg
  • 17 kg
  • 20 kg
Specifications
Product Information
  • Catalog #
    Supplier #
    Seq
    Pk
    Availability
    Price
  • Masticator Basic, 400ml, lacquered steel
    Masticator Basic, 400ml, lacquered steel
    710-0102
    470
    001
    1
    Log in for availability
    1 * 1 items
    1 * 1 items - 1 937,00 €
    1 937,00 €
    New Customer Promotional Price
    1 * 1 items
       1 937,00 €
    Specifications:
    More
    Accessories
    Specifications
    Model
    Classic/Basic
    Environmentally Preferable
    Motor speed
    1500 min⁻¹
    Reduction
    1:6
    Strokes
    8,5/s
    Timer
    15 - 90 seconds; digital setting, continuous mode
    Start/stop
    door activated
    Power
    270 W
    Motor
    Induction
    W×D×H
    240×400×260 mm
    Weight
    17 kg
  • Masticator Basic Panoramic
    Masticator Basic Panoramic
    710-0103
    475
    002
    1
    Log in for availability
    1 * 1 items
    1 * 1 items - 2 074,00 €
    2 074,00 €
    New Customer Promotional Price
    1 * 1 items
       2 074,00 €
    Specifications:
    More
    Accessories
    Specifications
    Model
    Classic/Basic
    Environmentally Preferable
    Motor speed
    1500 min⁻¹
    Reduction
    1:6
    Strokes
    8,5/s
    Timer
    15 - 90 seconds; digital setting, continuous mode
    Start/stop
    door activated
    Power
    270 W
    Motor
    Induction
    W×D×H
    240×400×260 mm
    Weight
    17 kg
  • Masticator Classic 400 ml, stainless steel
    Masticator Classic 400 ml, stainless steel
    710-0100
    400
    003
    1
    Log in for availability
    1 * 1 items
    1 * 1 items - 2 004,00 €
    2 004,00 €
    New Customer Promotional Price
    1 * 1 items
       2 004,00 €
    Specifications:
    More
    Accessories
    Specifications
    Model
    Classic/Basic
    Environmentally Preferable
    Motor speed
    1500 min⁻¹
    Reduction
    1:6
    Strokes
    8,5/s
    Timer
    15 - 90 seconds; digital setting, continuous mode
    Start/stop
    door activated
    Power
    270 W
    Motor
    Induction
    W×D×H
    240×400×260 mm
    Weight
    17 kg
  • Masticator Classic Panoramic
    Masticator Classic Panoramic
    710-0104
    500
    004
    1
    Log in for availability
    1 * 1 items
    1 * 1 items - 2 139,00 €
    2 139,00 €
    New Customer Promotional Price
    1 * 1 items
       2 139,00 €
    Specifications:
    More
    Accessories
    Specifications
    Model
    Classic/Basic
    Environmentally Preferable
    Motor speed
    1500 min⁻¹
    Reduction
    1:6
    Strokes
    8,5/s
    Timer
    15 - 90 seconds; digital setting, continuous mode
    Start/stop
    door activated
    Power
    270 W
    Motor
    Induction
    W×D×H
    240×400×260 mm
    Weight
    17 kg
  • Masticator Silver, 400 ml
    Masticator Silver, 400 ml
    710-0101
    420
    005
    1
    Log in for availability
    1 * 1 items
    1 * 1 items - 2 534,00 €
    2 534,00 €
    New Customer Promotional Price
    1 * 1 items
       2 534,00 €
    Specifications:
    More
    Accessories
    Specifications
    Model
    Silver
    Environmentally Preferable
    Motor speed
    500…1500 min⁻¹
    Reduction
    1:6
    Strokes
    5/6/7/8/s
    Timer
    5/30/60/90/120/150/180/210/240 s; keyboard digital setting, continuous mode
    Start/stop
    door activated
    Power
    270 W
    Motor
    Induction
    W×D×H
    240×400×300 mm
    Weight
    20 kg
  • Masticator Silver Panoramic
    Masticator Silver Panoramic
    710-0105
    520
    006
    1
    Log in for availability
    1 * 1 items
    1 * 1 items - 2 731,00 €
    2 731,00 €
    New Customer Promotional Price
    1 * 1 items
       2 731,00 €
    Specifications:
    More
    Accessories
    Specifications
    Model
    Silver
    Environmentally Preferable
    Motor speed
    500…1500 min⁻¹
    Reduction
    1:6
    Strokes
    5/6/7/8/s
    Timer
    5/30/60/90/120/150/180/210/240 s; keyboard digital setting, continuous mode
    Start/stop
    door activated
    Power
    270 W
    Motor
    Induction
    W×D×H
    240×400×300 mm
    Weight
    20 kg