You Searched For: Heating Mantles
24  results were found
SearchResultCount:"24"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: WINKLER GMBH
Description: For round bottomed flasks. Max. 450 °C. Multiposition heating mantles with powder coated, chemically resistant metal housing and stainless steel front...
Supplier: WINKLER GMBH
Description: For round bottomed flasks. Mantles have crocheted outer jacket made from glass silk (WHG2H models are made from silicate yarn). Sizes from 500 ml upwa...
Supplier: Cole-Parmer
Description: Provides powerful stirring speeds up to 520 min⁻¹ and heating up to 450 °C.
Supplier: Cole-Parmer
Description: Heating mantles with interchangeable cartridges.
Supplier: WINKLER GMBH
Description: Heating mantle, WHU2ER 20000, SS plug, Flask capacity: 20000 ml, 450 °C, Ø×H: 438×234 mm
Supplier: Cole-Parmer
Description: Flexible heating element for efficient heat transfer and fast boiling times.
Supplier: Cole-Parmer
Description: Magnetic stirrer and heating mantle in one convenient unit.
Supplier: Cole-Parmer
Description: Accommodate 60° funnels, pear-shaped flasks and round-bottom flasks.
Supplier: Cole-Parmer
Description: Thermal insulating cartridge provides maximum heat transfer.
Supplier: Cole-Parmer
Description: Designed to determine the protein content of food.
Supplier: Cole-Parmer
Description: Opening in base accommodates a wide variety of flasks and 60° funnels.
Supplier: WINKLER GMBH
Description: For round bottomed flasks. Max. 450 °C. Multiposition heating mantles with powder coated, chemically resistant metal housing and stainless steel front...
Catalog Number: (454-0045)
Supplier: Cole-Parmer
Description: Heating mantle, RE0250, Flask capacity: 250 ml
UOM: 1 * 1 items
Catalog Number: (ELET36013-14)
Supplier: Cole-Parmer
Description: Accepts 250 to 1000 ml round bottom flasks saving you money.
UOM: 1 * 1 items
Supplier: Cole-Parmer
Description: Don't get burned - the outside stays 'safe to the touch'.
Catalog Number: (WITGDH.WHM12018)
Supplier: WITEG LABORTECHNIK GMBH
Description: Heating mantle, Flask capacity: 6000 ml, Ø×H: 325×226 mm
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 24
no targeter for Bottom