You Searched For: Heat Sealers
12  results were found
SearchResultCount:"12"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (129-1160)
Supplier: VWR Collection
Description: Quickly and efficiently seal PP or PE bags, sheets or flat tubing from 0,01 to 0,8 mm thick.
UOM: 1 * 1 items
Supplier: MATRIX TECHNOLOGIES CORP. LTD
Description: ALPSâ„¢ 5000 is the next generation in plate sealing equipment offering fast sealing times, increased process customisation and all electric operation i...
Supplier: TUTTNAUER
Description: Impulse sealing unit for sealing of medical products. Sterilisation of medical instruments is a critically important activity in medicine and dentistr...
Supplier: MATRIX TECHNOLOGIES CORP. LTD
Description: Heat sealer, accessory, Plate carrier for non-skirted PCR plates, ALPSâ„¢
Catalog Number: (732-0996)
Supplier: MATRIX TECHNOLOGIES CORP. LTD
Description: Heat sealing offers superior sample protection of storage and reaction plates in applications including compound storage, sample archiving and PCR. Th...
UOM: 1 * 1 items
Catalog Number: (MATRAB-0400)
Supplier: MATRIX TECHNOLOGIES CORP. LTD
Description: Use of the ALPSâ„¢ 30 manual heat sealer protects samples from evaporation and contamination when performing PCR or sample storage. Perfect for sealing ...
UOM: 1 * 1 items
Supplier: EPPENDORF
Description: The Eppendorf HeatSealer Family offers reliable and effective sealing of a variety of plates. When working with plates, only a tight and reproducible ...
Catalog Number: (731-0352)
Supplier: Corning
Description: Axygen® PlateMax® semi automatic plate sealer is ideal for the low-to-medium throughput laboratory that requires uniform and consistent sealing of mic...
UOM: 1 * 1 items
Catalog Number: (SEVEFS3604)
Supplier: SEVERIN ELECTROGERAETE
Description: For vacuum sealing bags.
UOM: 1 * 1 items
Supplier: KOEHLER TECHNISCHE PRODUKTEN
Description: Heat sealer, bag, Electronic
Catalog Number: (MELA10010)
Supplier: MELAG Medizintechnik OHG
Description: MELAseal® Pro is a high performance rotary sealer for sealing transparent sterilisation packaging. The relevant sealing parameters, such as pressure, ...
UOM: 1 * 1 items
Supplier: HULME MARTIN HEAT SEALERS LTD
Description: Superseal Impulse heat sealers are suitable for a wide range of laboratory and medical applications. Units are used extensively for the protective pac...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 12 of 12
no targeter for Bottom