You Searched For: Shelves
77  results were found
SearchResultCount:"77"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Shelves, Stainless steel wire shelf, with slides set, 590×470 mm, max. shelf workload: 25 kg, For: IL 240PR/IL 240CR Premium cooled incubator
Supplier: VWR Collection
Description: Shelves, Stainless steel raised shelf, For: VWB2 26
Supplier: VWR Collection
Description: Shelves, Reinforced stainless steel wire shelf, with slides set, 590×470 mm, max. shelf workload: 100 kg, For: IL 240PR/IL 240CR Premium cooled incubator
Catalog Number: (390-0735)
Supplier: VWR Collection
Description: Shelves, Stainless steel shelf, For: INCU-Line® IL 150R/250R Premium
UOM: 1 * 1 items
Supplier: Metro International
Description: Simple to configure using a clip lock system - no tools required.
Sale
Supplier: Metro International
Description: The open wire design of these heavy-gauge shelves minimizes dust accumulation and allows a free circulation of air, greater visibility of stored items...
Supplier: Metro International
Description: The handles are compatible with both MetroMax i®and MetroMax Q® units to provide additional maneuverability and ergonomics.
Supplier: Metro International
Description: Designed to work in conjunction with the entire Super Erecta® system of shelves and accessories. These Super adjustable wire shelves have a unique cor...
Supplier: asecos
Description: Sturdy construction racks with four galvanized steel sumps shelves as storage levels and galvanized rack frames. Ideal for safe and approved storage o...
Supplier: Metro International
Description: Track sets includes necessary sections of track for assembling track runs up to 6400 mm. Only one track set is required between stationary units.
Supplier: Metro International
Description: The chrome finish mobile security standard-duty stem caster units protect valuable materials and sensitive items from loss and they are ideal for use ...
Supplier: MEMMERT
Description: Shelves, Perforated stainless steel shelf, For: IF260
Supplier: Metro International
Description: Shelves, Stationary post, SiteSelect™, chrome plated, 699 mm height, For: Drop Mat™ Super Erecta® display shelving, Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, APPEAL™ display shelving
Supplier: Metro International
Description: MetroMax iâ„¢ shelving offers lifelong durability - the strength of steel and a lifetime warranty against rust and corrosion. Removable polymer shelf ma...
Supplier: Metro International
Description: For stationary or mobile installations, ledges prevent items from protruding or falling from shelves.
Supplier: Metro International
Description: The stackable shelf ledges (51 mm) prevent items from protruding or falling from shelves.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 77
no targeter for Bottom