You Searched For: Furniture
291  results were found
SearchResultCount:"291"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Polished granite. Four absorbing feet dampen the transmission of extraneous vibrations to this slab making it suitable, not only for high precision we...
Supplier: VWR Collection
Description: Shelves, Reinforced stainless steel wire shelf, with slides set, 590×470 mm, max. shelf workload: 100 kg, For: IL 240PR/IL 240CR Premium cooled incubator
Catalog Number: (611-2549)
Supplier: VWR Collection
Description: High performance micro balances and analytical balances should be placed in an environment where interfering vibration is at a minimum, to give fast r...
UOM: 1 * 1 items
Catalog Number: (390-0735)
Supplier: VWR Collection
Description: Shelves, Stainless steel shelf, For: INCU-Line® IL 150R/250R Premium
UOM: 1 * 1 items
Supplier: VWR Collection
Description: Shelves, Stainless steel raised shelf, For: VWB2 26
Supplier: VWR Collection
Description: Shelves, Stainless steel wire shelf, with slides set, 590×470 mm, max. shelf workload: 25 kg, For: IL 240PR/IL 240CR Premium cooled incubator
Supplier: Metro International
Description: Designed to work in conjunction with the entire Super Erecta® System of shelves and accessories, these Super Adjustable wire shelves have a unique cor...
Supplier: Metro International
Description: Providing a complete shelving unit in a single box, Convenience Pakâ„¢ shelving is the easy-to-order alternative to separate shelving components.
...
Supplier: asecos
Description: Fireproof safety cabinets, classified type 30 (fire resistant up to 30 minutes), safe, durable, robust and convenient with transport base and bicolour...
Supplier: asecos
Description: Fireproof safety cabinets, classified type 90 (fire resistant up to 90 minutes).
Catalog Number: (MSGA30343001)
Supplier: asecos
Description: Safe and approved storage of aggressive and flammable hazardous substances in working areas.
UOM: 1 * 1 items
Supplier: asecos
Description: Fireproof safety cabinets, classified type 90 (fire resistant up to 90 minutes), safe and approved storage of aggressive and flammable hazardous subst...
Supplier: asecos
Description: Fireproof safety cabinets, classified type 90 (fire resistant up to 90 minutes), safe and approved storage of aggressive and flammable hazardous subst...
Supplier: asecos
Description: Fireproof safety cabinets, classified type 90 (fire resistant up to 90 minutes), safe, durable, robust and convenient with transport base and bicolour...
Supplier: asecos
Description: Fireproof safety cabinets, classified Type 90 (fire resistant up to 90 minutes), safe, durable, robust and convenient with transport base and bi-colou...
Supplier: asecos
Description: Under bench cabinets with drawers or wing doors. Fire resistance: 90 minutes.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 291
no targeter for Bottom