Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "extraction-cabinets"
https://si-bot.vwr.com/stibo/smallweb/std.lang.all/52/09/18945209.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"11"
  List View Grid View Easy View (new)
Sort by:
Supplier:  Bohlender
Description:   Anodized aluminium frame with panels made of polycarbonate.
Supplier:  AZBIL TELSTAR BENELUX B.V.
Description:   These powder fume hoods are used wherever harmful powders or dusts are released, e.g. when weighing. The hoods guarantee the highest degree of protect...
Supplier:  LABCONCO
Description:   In nanoparticle manipulation and dry powder chemical handling, enclosures keep hazardous powders and particulates contained. Room air is pulled into t...
Supplier:  BIGNEAT LTD.
Description:   Excel Plus cabinets are for precision weighing applications - for protection from particles whilst weighing and handling powders. These cabinets incor...
Supplier:  BIGNEAT LTD.
Description:   Cleantec LEV system is designed to solve fume, dust and odour problems. Efficient filtration within a powerful extraction system removes hazardous mat...
UOM:  1 * 1 items
Catalog Number: (LBCP3961622)
Supplier:  LABCONCO
Description:   Xpert® Bulk Powder Filtered System is designed to provide user protection from hazardous airborne particulates and fumes generated during powder trans...
UOM:  1 * 1 items
Supplier:  LABCONCO
Description:   XPert® balance enclosures provide user protection by keeping powders, particulates and fume contained during weighing procedures. They are well suited...
Catalog Number: (LBCP3960302)
Supplier:  LABCONCO
Description:   Balance enclosures, Xpert®, Balance enslosure, 3' XPert® Easy-Order, with guardian 1000 digital airflow monitor, Filter: +, W×D×H ext.: 914,4×736,6×726,4 mm, Weight: 83 kg
UOM:  1 * 1 items
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8  of 11
  1  2  Next