Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "biological-and-microbiological-safety-cabinets"
https://si-bot.vwr.com/stibo/smallweb/std.lang.all/49/45/18944945.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"20"
  List View Grid View Easy View (new)
Sort by:
Supplier:  LABCONCO
Description:   These Biosafety cabinets provide personnel, product and environmental protection from hazardous particulates such as agents that require Biosafety Lev...
Supplier:  AZBIL TELSTAR BENELUX B.V.
Description:   BioVanguard Green Line is a Class II cabinet range which offers simplicity, robustness and high reliability and ensures the highest level of protectio...
Supplier:  LABCONCO
Description:   The cabinet provides personnel, product and environmental protection and is suitable for work in biosafety levels 1, 2 and 3. These biohazard cabinets...
Supplier:  AZBIL TELSTAR TECHNOLOGIES S.L
Description:   The Bio II Advance has been developed to maximise the convenience, comfort and safety for the user, the sample and the environment whilst minimising t...
Supplier:  LABCONCO
Description:   Purifier Class I enclosures protect the user from agents that require biosafety level 1, 2 or 3 containment but no product protection.
Supplier:  AZBIL TELSTAR BENELUX B.V.
Description:   BioVanguard Greenline B is a class II cabinet range which offers simplicity, robustness and high reliability and ensures ensures the highest level of ...
Supplier:  LABCONCO
Description:   Biosafety cabinets, Purifier® Logic®, Class II, Type A2, nominal width 4 feet, with UV light, service fixtures and 8” sash opening, Schuko plug, Exhaust air volume (m³/h): 457,27 - 503,17, External W×D×H (mm): 1379,22×792,48×1559,56, Weight (kg): 265
Supplier:  LABCONCO
Description:   Purifier HEPA filtered enclosures protect the user from agents that require biosafety level 1, 2 or 3 containment but no product protection. With ergo...
UOM:  1 * 1 items
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8  of 20
  1  2  3  Next