Chest freezers, HERAfreeze™ HFC
Supplier: Thermo Fisher Scientific
HERAfreeze®
471-1444EA 0 EUR
471-1444 471-1449
Chest freezers, HERAfreeze™ HFC
Freezers Cryogenic Freezers
An alternative to storage in liquid nitrogen. Product protection by means of low application temperature, even with higher ambient temperatures (+16 to +32°C, temperature class N).
  • Uniform temperature
  • Cover with counterbalance and a silicon, magnetic seal and insulated intermediate lid
  • Patented compression-type refrigeration system, long service life
  • Intuitive operation and easy-to-understand symbols
All devices come with transport rollers.
Inner housing made from corrosion-resistant stainless steel.
High pressure polyurethane foam insulation contains no CFCs, HCFCs or HFCs.
Wide range of accessories for sample storage and management.
Temperature: −140 °C (HFC 10140)/−150 °C (HFC 7150).