Standard chest freezers
Supplier: Liebherr
471-0287EA 739 EUR
471-0287 471-0288 LIEBGTL6106 471-0925 471-1022 471-0286
Standard chest freezers
Freezers Laboratory / General Purpose Freezers
Commerical chest freezers featuring StopFrost technology to reduce the build up of ice.
  • Analogue display
  • Manual defrosting
Temperature range: −14 to −26 °C.