You Searched For: Automatic Extractor Systems
7  results were found
SearchResultCount:"7"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (GILI26150006)
Supplier: GILSON LCMS
Description: This compact ASPEC® system combines positive pressure solid phase extraction (SPE) with small footprint and precise liquid handling for reliable autom...
UOM: 1 * 1 items
Catalog Number: (GILI2614010)
Supplier: GILSON LCMS
Description: This ASPEC® system for positive pressure solid phase extraction (SPE) automation maximises throughput by processing up to four samples in parallel. Wi...
UOM: 1 * 1 items
Catalog Number: (VELPF30520204)
Supplier: VELP SCIENTIFIC
Description: In order to perform a reliable raw fibre analysis, the sample must have a low fat content (\<1%). For those samples that exceed this value, a prelimin...
UOM: 1 * 1 items
Supplier: VELP SCIENTIFIC
Description: A fully automated and cloud enabled solvent extractor capable of a high sample throughput, offering state-of-the-art technology for a fast, precise an...
Supplier: VELP SCIENTIFIC
Description: The SER 148/3 and SER 148/6 can be used to separate a substance or a group of elements (e.g. fat) from solid and semi-solid samples according to the R...
Supplier: GERHARDT
Description: SOXTHERM® units are automatic and fully programmable extraction equipment for the determination of fat in foodstuffs, including dairy, cereals, fruit,...
Supplier: BEHR
Description: These extractors can be used to separate a substance or a group of elements from samples according to the Randall technique. It features lever assembl...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 7 of 7
no targeter for Bottom