You Searched For: Extractors
71  results were found
SearchResultCount:"71"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Pure cellulose cotton. Suitable for all types of extractors (Soxhlet, Büchi, Gerhardt and more).
Supplier: VWR Collection
Description: Borosilicate 3.3 glass.
Supplier: Lenz Laborglas GmbH & CO.KG
Description: DURAN®, Borosilicate glass.
Supplier: Lenz Laborglas GmbH & CO.KG
Description: DURAN®, borosilicate glass, extractor head only.
Catalog Number: (GILI2614010)
Supplier: GILSON LCMS
Description: This ASPEC® system for positive pressure solid phase extraction (SPE) automation maximises throughput by processing up to four samples in parallel. Wi...
UOM: 1 * 1 items
Supplier: Whatman products (Cytiva)
Description: Grade 603 standard cellulose extraction thimbles, made of high quality cellulose, are available in a range of sizes suitable for use in thimble holder...
Supplier: Whatman products (Cytiva)
Description: Cytiva's Whatman cellulose thimbles are manufactured from high-quality alpha cellulose cotton linter for high-performance use in automated Soxhlet ext...
Supplier: Whatman products (Cytiva)
Description: Cytiva's Whatman 603G borosilicate glass fiber thimbles with inorganic binder are flexibly designed for use in a full range of commercially available ...
Supplier: Lenz Laborglas GmbH & CO.KG
Description: DURAN®, Borosilicate glass.
Catalog Number: (LENZ05367073)
Supplier: Lenz Laborglas GmbH & CO.KG
Description: Soxhlet Extractor, Complete with Dimroth condenser and flask, LF 100 flange, Extraction cap.: 2000 ml, Flask capacity: 2000 ml, Socket size: 45/40 NS
UOM: 1 * 1 items
Supplier: Lenz Laborglas GmbH & CO.KG
Description: DURAN®, Borosilicate glass.
Supplier: Lenz Laborglas GmbH & CO.KG
Description: DURAN®, borosilicate glass. For extraction from solids with solvents.
Supplier: Lenz Laborglas GmbH & CO.KG
Description: DURAN®, borosilicate glass.
Supplier: Lenz Laborglas GmbH & CO.KG
Description: DURAN®, borosilicate glass.
Supplier: ROBU GLASFILTER
Description: Borosilicate glass 3.3.
Catalog Number: (537-0028)
Supplier: Whatman products (Cytiva)
Description: High-purity Whatman quartz thimbles from Cytiva support accurate, reproducible emission testing by providing low extractable levels and temperature re...
UOM: 1 * 10 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 71
no targeter for Bottom