Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "ion-meters"
https://si.vwr.com/stibo/smallweb/std.lang.all/40/53/18374053.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"10"
  List View Grid View Easy View (new)
Sort by:
Supplier:  HORIBA
Description:   These pocket meters directly measure nitrate ion and nirate-nitrogen concentrations in 0,3 ml sample (or 0,05 ml sample with sampling sheet B). The un...
Supplier:  HORIBA
Description:   These waterproof pocket meters directly measure calcium, potassium or sodium concentrations in 0,3 ml sample (or 0,05 ml sample with sampling sheet B)...
Supplier:  HORIBA
Description:   These waterproof pocket meters provide direct measurements of different ions in liquid, solid, viscous or powder samples using a unique flat sensor wi...
Catalog Number: (HANNHI93734)
Supplier:  Hanna
Description:   HI 93734 meter measures the total and free chlorine (Cl2) content in water and wastewater.
UOM:  1 * 1 items
Discontinued
Catalog Number: (HANNHI93717)
Supplier:  Hanna
Description:   The HI 93717 meter measures the phosphate (PO43-) content in water and wastewater in the 0,0 to 30,0 mg/l (ppm) range. The meter...
UOM:  1 * 1 items
Discontinued
Catalog Number: (HANNHI93702)
Supplier:  Hanna
Description:   The HI 93702 meter measures the copper content in water and wastewater in the 0,00 to 5,00 mg/l (ppm) range. The meter uses an exclusive positive-lock...
UOM:  1 * 1 items
Discontinued
Supplier:  Hanna
Description:   The HI 98202 is an easy to use and inexpensive instrument for determining water hardness or softness.
UOM:  1 * 1 items
Supplier:  Hanna
Description:   Total hardness, that is the presence of magnesium and calcium, is due mainly to the run-off water dissolving these salts as it flows or filters throug...
UOM:  1 * 1 items
Discontinued
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8  of 10
  1  2  Next