You Searched For: Freeze Dryers
11  results were found
SearchResultCount:"11"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: LABCONCO
Description: FreeZone® console freeze dryers with or without stoppering tray dryers are designed for lyophilising moderate to large loads and stoppering vials or s...
Safer Human and Environmental Health
Sustainable Materials
Sustainable Packaging
Supplier: LABCONCO
Description: FreeZone® bench top freeze dryers are designed for lyophilising light to moderate sample loads. The compact design is ideal for laboratories with limi...
Safer Human and Environmental Health
Sustainable Materials
Sustainable Packaging
Supplier: LABCONCO
Description: Designed to handle the lyophilisation needs of research laboratories. Collectors that reach −50 °C are available with capacities from 1 to 18 litres. ...
Catalog Number: (LBCP780701030)
Supplier: LABCONCO
Description: These FreeZone® small tray dryers provide precise control and program flexibility for consistent performance and are designed for lyophilising small t...
UOM: 1 * 1 items
Safer Human and Environmental Health
Sustainable Materials
Sustainable Packaging
Catalog Number: (535-0412)
Supplier: LABCONCO
Description: The FreeZone® Triad™ freeze drying system is an all inclusive bench top lyophiliser for 4 samples on the left side and stoppering tray dryer processin...
UOM: 1 * 1 items
Catalog Number: (LBCP7522700)
Supplier: LABCONCO
Description: Benchtop freeze drying system, type 304 stainless steel with 12 ports, -75 °C. Ideal for the laboratory with occasional lyophilisation requirements, t...
UOM: 1 * 1 items
Catalog Number: (CHRS102141)
Supplier: CHRIST, MARTIN GMBH & CO KG
Description: Freeze dryers are ideal for effective lyophilisation of specimens with wireless shelf technology. Compact tabletop unit for effective lyophilisation o...
UOM: 1 * 1 items
Catalog Number: (CHRS102370-1)
Supplier: CHRIST, MARTIN GMBH & CO KG
Description: Alpha 1-4 LSC Basic Universal package in a 'universal configuration' with RV8 Edwards vacuum pump. The package contains everything you need to use.
UOM: 1 * 2 items
Supplier: CHRIST, MARTIN GMBH & CO KG
Description: LDplus (Lyo Display Plus) stands for a comfortable, easy-to-use and self-explaining user interface for controlling routine freeze drying processes, co...
Catalog Number: (CHRS102141-1)
Supplier: CHRIST, MARTIN GMBH & CO KG
Description: The Alpha 1-4 LSCplus universal package with vacuum pump RV8. The freeze-drying package includes everything you need to use.
UOM: 1 * 2 items
Catalog Number: (CHRS101252-1)
Supplier: CHRIST, MARTIN GMBH & CO KG
Description: RVC 2-25 CDplus universal package with vacuum chemistry diaphragm pump MZ 2C includes everything you need to use.
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 11 of 11
no targeter for Bottom