You Searched For: Distillation Systems
15  results were found
SearchResultCount:"15"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VELP SCIENTIFIC
Description: The UDK Series distillers are designed to meet the most challenging demands and requirements for diverse applications, according to international stan...
Catalog Number: (VELPF307A0660)
Supplier: VELP SCIENTIFIC
Description: Designed to neutralise toxic fumes produced during mineralisation and safeguard lab operators without requiring any connection to tap water.
UOM: 1 * 1 items
New Product
Catalog Number: (709-0140)
Supplier: VELP SCIENTIFIC
Description: The UDK 169 distillation and titration system is designed to perform nitrogen and protein determination according to the Kjeldahl Method (TKN) in the ...
UOM: 1 * 1 items
Supplier: BUCHI
Description: A modular, upgradeable, system for scrubbing of harmful fumes from distillation or digestion processes. The system can be obtained with up to four pro...
Supplier: BEHR
Description: Distillation apparatus for determining the alcohol content and the volatile acids in wine and other alcoholic drinks.
Supplier: BUCHI
Description: KjelMaster K-375 provides solutions designed for convenient nitrogen and protein analysis.
Catalog Number: (WITG2.530.000)
Supplier: WITEG LABORTECHNIK GMBH
Description: Kjeldahl distillation unit made of borosilicate glass.
UOM: 1 * 1 items
Supplier: BEHR
Description: The behrotest® WE 5 Five-in-a-line multiple steam generator provides the simultaneously rapid and gentle distillation of up to five samples. The disch...
Supplier: GERHARDT
Description: Distillation system, Kjeldahl, Manual, KI 12/26, Array: 6, Flask capacity: 750 ml
Catalog Number: (705-0857)
Supplier: GERHARDT
Description: VAPODEST® 200 is the ideal instrument for distillation of volatiles, SO₂, alcohol and phenol.
UOM: 1 * 1 items
Supplier: BEHR
Description: behr steam distillation units are not just high performance units and reliable partners in your daily laboratory routine. An essential factor in the d...
Supplier: GERHARDT
Description: The VAPODEST® 400 and 450 are designed with extended funtionality to allow easy, comfortable operation of Kjeldahl analysis.
Supplier: GERHARDT
Description: Two level apparatus for six mineralisations and six distillations.
Catalog Number: (705-0858)
Supplier: GERHARDT
Description: VAPODEST® 300 is the ideal rapid water steam distllation instrument for Kjeldahl distillation. All glass components involved in the distillation proce...
UOM: 1 * 1 items
Catalog Number: (705-0488)
Supplier: GERHARDT
Description: The Turbosog scubber unit condenses and neutralises aggressive acid vapours from the Kjeldahl digestion, including perchloric acid and other toxic gas...
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 15 of 15
no targeter for Bottom