You Searched For: Dilutors
9  results were found
SearchResultCount:"9"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Gravimetric dilutors are used for the dilution of a sample before microbiological analysis.
Supplier: VWR Collection
Description: Gravimetric dilutors are used for the dilution of a sample before microbiological analysis. Robotic arm with the 5 kg model features easy sample dropp...
Sale
Supplier: HAMILTON BONADUZ
Description: The Microlab® 600 dual syringe diluter is a semi-automated liquid handling instrument capable of dilutions up to 1:50000 in a single step. The precisi...
Supplier: INTERSCIENCE
Description: DiluFlow® Elite automatically dilutes in seconds a solid sample with the appropriate weight of diluent with full traceability. Programmable and...
Supplier: INTERSCIENCE
Description: DiluFlow® is a gravimetric dilutor for the dilution of the sample before its microbiological analysis.
Supplier: INTERSCIENCE
Description: DiluFlow® Pro automatically dilutes in seconds a solid sample with the appropriate weight of diluent with traceability. With its robotic arm an...
Supplier: IUL Instruments SA
Description: The Smart Dilutor W automates and facilitates the precise gravimetric dilution of samples, that is a key process stage in microbiology laboratories.Th...
Catalog Number: (710-0862)
Supplier: IUL Instruments SA
Description: Equipment specifically designed to facilitate the controlled addition of media into a homogeniser/lab blender bag using a load cell. The Pinch Dilutor...
UOM: 1 * 1 items
Catalog Number: (GOSSS-DILUTER-1)
Supplier: Corning
Description: The Corning® Gosselin™ S-Diluter-1 is an automated food sample diluter suitable for the analysis of a range of sample sizes from 50 to 400 ml.
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 9 of 9
no targeter for Bottom