You Searched For: Digestion Systems
43  results were found
SearchResultCount:"43"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Chemicals
Description: Kjeldahl tablets are catalysts that promote the digestion step of the Kjeldahl method for nitrogen determination.
Catalog Number: (330666A)
Supplier: VWR Chemicals
Description: This product is not available in all countries. Please check with your local Avantor International office or supplier.
UOM: 1 * 1.000 Tablet
Catalog Number: (ACRO243920025)
Supplier: Thermo Scientific
Description: Kjeldahl tablets with selenium (Wieninger), 5 g, Contents: Na₂SO₄: 4875 mg, Cu(II)SO₄.5H₂O: 75 mg; Se: 50 mg, Ø: 18 - 20 mm
UOM: 1 * 2,5 kg
MSDS
Supplier: Bohlender
Description: Dimensionally stable basic vessel and screw cover made of TFM, homogenous, non porous surface.
Supplier: VELP SCIENTIFIC
Description: A series of thermoreactors for digestion of samples for a range of analyses including COD, heavy metal, non metallic elements. The ECO 6 has the possi...
Supplier: VELP SCIENTIFIC
Description: Fully automatic Kjeldahl digestion units for nitrogen analysis and protein determination that incorporate the TEMSâ„¢ technology for savings in terms of...
Supplier: VELP SCIENTIFIC
Description: DK digesters are traditional digesters, consisting of an aluminium heating block that offers excellent thermal homogeneity, heating up to 450 °C, prov...
Supplier: THOMPSON & CAPPER
Description: Kjeldahl catalyst reagents in a convenient tablet form with a range of tablets to suit general and specific applications.
Supplier: BUCHI
Description: Digestion of protein samples with sulphuric acid and catalyst requires prolonged periods of heating and is traditionally carried out in block heaters....
Catalog Number: (HOFM40061.00)
Supplier: HOFMANN GLASTECHNIK GMBH
Description: Arsenic limit test apparatus, complete
UOM: 1 * 1 items
Catalog Number: (HOFM40065.00)
Supplier: HOFMANN GLASTECHNIK GMBH
Description: This apparatus is suitable for the specification of arsenic.
UOM: 1 * 1 SET
Catalog Number: (BEHRB00217639)
Supplier: BEHR
Description: In the behrotest® nitrogen station, solvent is evaporated under ambient temperature conditions by using a mild nitrogen flow. A distributor leads the ...
UOM: 1 * 1 items
Catalog Number: (BEHRB00217638)
Supplier: BEHR
Description: The complete sample clean up station includes dropping funnel, clean-up column with glass frit, reducing flasks and stand.
UOM: 1 * 1 items
Catalog Number: (BUCH1154461000)
Supplier: BUCHI
Description: The KjelDigester provides fast and safe digestion of up to 20 samples simultaneously.
UOM: 1 * 1 items
Supplier: GERHARDT
Description: Traditional Kjeldahl digestion units for flask sizes from 50 to 750 ml.
Catalog Number: (PARR4744)
Supplier: PARR
Description: The system is designed specifically for preparing laboratory samples for chemical analysis.
UOM: 1 * 1 Each
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 43
no targeter for Bottom