Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • In Stock (3)
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
density-meters
https://si.vwr.com/stibo/smallweb/std.lang.all/51/02/18945102.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"6"
  List View Grid View Easy View (new)
Sort by:
Catalog Number: (614-1595)
Supplier:  METTLER / TOLEDO
Description:   Suitable for measuring the densities of liquids, this instrument uses the oscillating body method.
UOM:  1 * 1 items
Catalog Number: (SIMELD0560)
Supplier:  TQC
Description:   This densimeter is designed to perform automatic calculation of density and compensation of water temperature.
UOM:  1 * 1 items
Catalog Number: (634-0882)
Supplier:  BIOCHROM (SCIE-PLAS LTD)
Description:   The Ultrospec 10 Classic can measure growth rates of E. coli, other bacteria and yeast in suspension at 600 nm.
UOM:  1 * 1 KIT
Supplier:  GRANT INSTRUMENTS LTD.
Description:   Densitometers for turbidity of cell suspensions.
Catalog Number: (PAARDMA500)
Supplier:  Anton Paar
Description:   This density meter provides a self-explanatory user interface to operate the instrument with one hand in a few moments.
UOM:  1 * 1 items
Catalog Number: (PAAR94680)
Supplier:  Anton Paar
Description:   DMA 500 is a laboratory density meter providing measurement with quality.
UOM:  1 * 1 items
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 6  of 6