You Searched For: Temperature Control Systems
9  results were found
SearchResultCount:"9"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: JULABO GmbH
Description: PRESTO® systems cover a working temperature range of −93 to +250 °C with high cooling and heating capacity. These instruments can compensate for exoth...
Catalog Number: (JULA9021709.D)
Supplier: JULABO GmbH
Description: The JULABO beer forcing test bath in conjunction with a photometer determines the product life of beer before clouding. The simulated aging process is...
UOM: 1 * 1 items
Supplier: LAUDA
Description: Process thermostats with integrated cooling system for dynamic temperature control within external circuits.
Supplier: LAUDA
Description: The LOOP product range includes two thermo-electric circulation thermostats for external temperature control.
Supplier: HUBER
Description: Unistats® are the ideal solution for fast and precise thermal control of externally connected applications. These compact units enable previously unac...
Supplier: IKA
Description: ICC compact immersion circulators allow for flexible coupling to water baths of different volumes. They are designed for tempering liquids up to tempe...
Supplier: HUBER
Description: Unistat® technology guarantees precise, reproducible temperature control results, short heating and cooling times and large temperature ranges without...
Catalog Number: (HUBE2041.0001.01)
Supplier: HUBER
Description: BFT5 is a special heating and cooling thermostat ideal for beer-forcing test. This unit is fitted with True Adaptive temperature control and programme...
UOM: 1 * 1 items
Catalog Number: (IKAA25004524)
Supplier: IKA
Description: This compact refrigerated and heating circulator HRC 2 control is well known for its high energy efficiency and a powerful and infinitely variable pre...
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 9 of 9
no targeter for Bottom