You Searched For: Heating Circulators
9  results were found
SearchResultCount:"9"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (473-0031)
Supplier: VWR Collection
Description: Small, low cost recirculator that handles applications such as thawing plasma and tempering photographic solutions. The sealed reservoir can control c...
UOM: 1 * 1 items
Supplier: JULABO GmbH
Description: High temperature circulators can be used for temperature applications up to 400 Â°C; units provide powerful heating capacities for shortest heat-u...
Save money with energy-efficient circulators!
Partial load operation is sufficient for most lab temperature control processes. Save on operating costs NOW while reducing your environmental impact.
ORDER TODAY
Go Green - Natural refrigerants
Trust +50 years experience
Supplier: JULABO GmbH
Description: The powerful and robust CORIOâ„¢ laboratory circulators provide the exact temperature, absolute precision, and a wide working temperature range. The COR...
Supplier: LAUDA
Description: The PRO heating bath thermostats are optimised for internal bath applications and offer excellent temperature stability. The removable operating units...
Supplier: LAUDA
Description: The PRO heating circulation thermostats are designed for external applications up to 250 °C. The low filling volume enables rapid heating in external ...
Supplier: HUBER
Description: These compact hotbox heating circulators are ideal for installation in production systems. All models have self-optimising True Adaptive Control for t...
Supplier: JUCHHEIM Laborgerate GmbH
Description: These thermostats have a special design that ensures trouble-free continuous operation at temperatures up to 350 °C. They are built in stainless steel...
Catalog Number: (635-2650)
Supplier: A.KRÃœSS Optronic
Description: A versatile compact high-performance instrument with electronic Peltier thermostat that can be used for controlling refractometer and viscometer tempe...
UOM: 1 * 1 items
Catalog Number: (HAAK222-1937)
Supplier: HAAKE
Description: SC100- A10 refrigerated bath circulator provides efficient cooling for open and closed applications, with temperatures ranging from −10 to +100°C and ...
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 9 of 9
no targeter for Bottom