You Searched For: Closed Circulator Systems
60  results were found
SearchResultCount:"60"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: These circulating chillers are an environmentally friendly, water-conserving and reliable means of controlling the temperature of liquids in open tank...
Water Efficient
Supplier: VWR Collection
Description: Stainless steel circulating baths with choice of Advanced Programmable or Advanced Digital temperature controller. Both are easy to navigate with larg...
Supplier: VWR Collection
Description: Stainless steel baths with user friendly MX temperature controller plus three control buttons and on-screen prompts. Baths can be used for external cl...
Catalog Number: (473-0031)
Supplier: VWR Collection
Description: Small, low cost recirculator that handles applications such as thawing plasma and tempering photographic solutions. The sealed reservoir can control c...
UOM: 1 * 1 items
Supplier: VWR Collection
Description: VWR® recirculating chillers are ideal for use with rotary evaporators, as they ensure constant, reproducible cooling conditions and avoid wasting tap ...
Supplier: JULABO GmbH
Description: Environmentally friendly recirculating coolers that saves tap water, these units were designed for simple cooling tasks where reliable continuous oper...
Water Efficient
Supplier: JULABO GmbH
Description: Environmentally friendly recirculating coolers that save tap water, units are rapid and powerful, and offer short cool-down times and high efficiency....
Energy Efficient
Water Efficient
Supplier: JULABO GmbH
Description: High temperature circulators can be used for temperature applications up to 400 Â°C; units provide powerful heating capacities for shortest heat-u...
Catalog Number: (461-3170)
Supplier: JULABO GmbH
Description: FD flow through coolers are used for cooling a loop circuit of a closed system, the unit must be connected in the return line of a heating circulator....
UOM: 1 * 1 items
Supplier: JULABO GmbH
Description: Compact, ergonomically designed chillers which have integrated stainless steel tanks with large volumes. Models can be either air or water cooled. App...
Energy Efficient
Water Efficient
Catalog Number: (QSON4905-220)
Supplier: QSONICA LLC
Description: Sonication generates heat which may be detrimental to some applications. Attempting to control temperature with ice and/or repeatedly changing out wat...
UOM: 1 * 1 items
Supplier: JULABO GmbH
Description: Recirculating coolers can handle virtually any cooling requirements in laboratories or industrial environments efficiently.
Catalog Number: (SCOI285414320)
Supplier: SI Analytics
Description: The flow-through cooler CK 310 is the perfect complement to bath thermostats of CT 72 series to extending their temperature range to below ambient tem...
UOM: 1 * 1 items
Supplier: JULABO GmbH
Description: PRESTO® systems cover a working temperature range of −93 to +250 °C with high cooling and heating capacity. These instruments can compensate for exoth...
Supplier: JULABO GmbH
Description: Ultra-low temperature refrigerated circulators for heating and cooling. The units are suitable for external temperature applications with simultaneous...
Supplier: JULABO GmbH
Description: FT immersion coolers are primarily used for counter cooling when connected to a heating circulator, or for rapidly cooling fluids down to low temperat...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 60
no targeter for Bottom