Temperature and humidity chart recorders, TH6
Supplier: Dickson
620-2504EA 1540 EUR
620-2504
Temperature and humidity chart recorders, TH6
Chart Recorders Recorders
The TH6 chart recorders will accommodate a wide range of temperatures and humidities. They feature a Ø 152 mm circular chart smartly housed in a compact bench mount with a transparent front panel. Recording time is user-selectable: 24 hours or 7 days.
  • °C/°F switchable
  • Panel mountable, DIN sized enclosure
  • Recording times, ranges and temperature values are user-selectable
Delivery information: Supplied with AC power supply, battery back-up with power status indicator, red and blue pens plus a chart.