Calorimeters, C 2000 basic and C 2000 control
Supplier: IKA
622-0025EA 0 EUR
622-0025 622-0024 622-0040 622-0041
Calorimeters, C 2000 basic and C 2000 control
Calorimeters
C 2000 basic and C 2000 control calorimeters are compact, modular systems designed to determine the calorific power of liquid and solid samples. They are characterised by a high degree of automation combined with exceptional ease of use.
  • Thermostatisation, automatic filling and emptying of the calorimeter's reservoirs
  • Calorimetric bombs are identified and filled with oxygen automatically
  • Cold water is supplied via a connection to a KV 600 thermostat or a fixed connection (C 25 pressure control valve* recommended)
  • C 5010 standard calorimetric bomb is resistant to halogens and can be mounted on a C 14 disposable crucible*
  • Connection interface for balance, printer, monitor and IKA C 5020 sample holder*; option to connect LIMS
The user can choose between two measuring modes: isoperibolic and dynamic.
Certifikace: Certified GHOST, DIN 51900, ISO 1928, ASTM D240, ASTM D4809, ASTM D5865, ASTM D1989, ASTM D5468 and ASTM E711.
Informace pro objednávání: C 2000 basic calorimeters, version 1 and C 2000 control calorimeters, version 1 are supplied with C 5010 standard calorimetric bomb; C 2000 basic version 2 is supplied with halogen-resistant calorimetric bombs C 5012, and C 5040 CalWin calorimetry software. * Not included - For details of the accessories, please contact your local Avantor sales office.