You Searched For: Calorimeters
6  results were found
SearchResultCount:"6"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (DIDAPTD039501)
Supplier: DIDALAB
Description: This calorimeter includes an aluminium container of 500 ml, concentric to one large neck Dewar vase. The metal internal vase is supported by plastic r...
UOM: 1 * 1 items
Catalog Number: (PARR6200EF)
Supplier: PARR
Description: Model 6200 is a microprocessor controlled, isoperibol oxygen bomb calorimeter which is widely used for both routine and occasional calorific tests.
UOM: 1 * 1 items
Catalog Number: (458-0052)
Supplier: IKA
Description: The AOD 1 is used for oxidative decomposition of organic samples under pressure in a closed system.
UOM: 1 * 1 items
Supplier: IKA
Description: The C 7000 calorimeter is designed to measure the calorific power of samples in a dry procedure.
Catalog Number: (IKAA10002387)
Supplier: IKA
Description: Compact and space-saving semi automated calorimeter to determine calorific values of liquid and solid samples.
UOM: 1 * 1 items
Catalog Number: (PARR1341EE)
Supplier: PARR
Description: Plain oxygen calorimeter, 230 V, 50/60 Hz
UOM: 1 * 1 Each
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 6 of 6
no targeter for Bottom