Dot and slot blotters
700-7245EA 668 EUR
VWRI700-7245 VWRI700-7094
Dot and slot blotters
Machined from high density acrylic with precision lapped mating surfaces to ensure uniform filter contact, these manifolds are designed for DNA and RNA membrane hybridisations and antigen/antibody screening in immunological applications. Each manifold consists of paired upper dot- or slot-forming modules and a lower vacuum chamber module. A vacuum of approximately 600 mm Hg (0,8 bar) is required during sample application.
  • Simple construction, easy to assemble by aligning the mating screws and tightening the stainless steel thumbscrews
  • Marked with an alphanumeric grid for sample identification