You Searched For: Blocks
32  results were found
SearchResultCount:"32"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Block heater block, VWR®, Single block, Well size: Ø 28,8 mm, For: 28 mm vials, No. of holes: 6, Depth: 45,0 mm
Supplier: VWR Peqlab
Description: Uniformly molded high-grade aluminum blocks provide excellent temperature accuracy.
Sale
Supplier: VWR Collection
Description: Solid blocks: For use as a low temperature hotplate or for custom drilling.
Supplier: VWR Collection
Description: Block heater block, VWR®, Mini block without thermometer well, Well size: Ø 6,4 mm, For: 0,2 ml tubes, No. of holes: 40, Depth: 15,2 mm
Supplier: VWR Collection
Description: Block heater block, VWR®, Aluminium block, For: 0,5 and 1,5 ml tubes, No. of holes: 26 + 24
Supplier: VWR Collection
Description: Block heater block, VWR®, Interchangeable tube block, Well size: Ø 11,1 mm, For: 24×1,5 ml tubes**, No. of holes: 24, Depth: 33,5 mm
Supplier: VWR Collection
Description: Constructed from a solid anodised aluminium block.
Supplier: GRANT INSTRUMENTS LTD.
Description: Grant blocks have a depth of 63 mm, ensuring precise temperature control for test tube applications.
Supplier: Cole-Parmer
Description: Interchangeable heating blocks offer excellent thermal conductivity.
Supplier: GRANT INSTRUMENTS LTD.
Description: Block heater block, Grant-bio, Block, interchangeable for 10×Ø 16 mm tubes
Supplier: IKA
Description: Block heater block, Single block, 30×8,3 mm Ø holes, For: Tubes, 6 mm, with round bottom, Depth: 48,4 mm
Catalog Number: (NABE602211388)
Supplier: NABERTHERM GMBH
Description: Heating elements for N7 (1 Set).
UOM: 1 * 1 SET
Catalog Number: (460-0049)
Supplier: Cole-Parmer
Description: Made from anodised aluminium, blocks have a separate hole to accommodate a thermometer if required.
UOM: 1 * 1 items
Supplier: GRANT INSTRUMENTS LTD.
Description: Block heater block, Grant-bio, Block for TS-DW, For: One Sarstedt® 96/2200 µl deep well plate
Supplier: BARNSTEAD (INC THERMOLYNE)
Description: Blocks for digital block heater to optimise flexibility and performance.
Supplier: IKA
Description: Reaction blocks allow syntheses to be carried out in round-bottomed flasks at temperatures up to 180 °C, avoiding the use of oil baths. The system ens...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 32
no targeter for Bottom