You Searched For: Block Heater/Coolers
10  results were found
SearchResultCount:"10"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (GRANCH3-150EU)
Supplier: GRANT INSTRUMENTS LTD.
Description: CH3-150, dry-block thermostat with heating and cooling function, is designed to thermally stabilise materials at a wide range of temperatures. With ch...
UOM: 1 * 1 items
Supplier: GRANT INSTRUMENTS LTD.
Description: The Grant bio block heater/coolers are compact, flexible and easy to use – they are ideal for the rapid heating and cooling of microtubes, and a very ...
Catalog Number: (TECN24120-00)
Supplier: Cole-Parmer
Description: Keep samples cold, clean, and dry.
UOM: 1 * 1 items
Catalog Number: (460-0226)
Supplier: EPPENDORF
Description: ThermoStatâ„¢ C is ideal for accurately setting and maintaining temperatures, Peltier technology provides precise temperature control. A digital display...
UOM: 1 * 1 items
Catalog Number: (BARN88880030)
Supplier: BARNSTEAD (INC THERMOLYNE)
Description: Compact programmable heating/cooling block heater with digital display can be used for all common applications in laboratory. Applications include enz...
UOM: 1 * 1 items
Catalog Number: (BOEI115001-2)
Supplier: BOEKEL SCIENTIFIC
Description: Boekel scientific's dry block heaters are advanced, configurable and easy-to-use.
UOM: 1 * 1 items
New Product
Supplier: HAEP LABOR CONSULT
Description: Block Thermostats for two blocks featuring high temperature accuracy, rapid heating times and even temperature distribution. The TH 21 unit features E...
Supplier: ASYNT
Description: For controlled cooling of 25 to 1000 ml flasks without using jacketed reaction vessels. Units allow active temperature control from +150 to −30 °C (de...
Catalog Number: (HACHLTV073)
Supplier: Hach
Description: Block heater/cooler, Hach®, LT 20, Thermostatic block heater, bench-top, 80 % relative humidity, non-condensing, Temperature range: 10...30 °C
UOM: 1 * 1 items
Catalog Number: (TECN36620-17)
Supplier: Cole-Parmer
Description: Safely maintain biological samples.
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 10 of 10
no targeter for Bottom