You Searched For: Block Heaters
80  results were found
SearchResultCount:"80"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Peqlab
Description: Uniformly molded high-grade aluminum blocks provide excellent temperature accuracy.
Sale
Supplier: VWR Collection
Description: Constructed from a solid anodised aluminium block.
Supplier: VWR Collection
Description: Block heater block, VWR®, Mini block with thermometer well, not suitable for use with mini block heaters with heated lid, Well size: Ø 30,0 mm, For: 50 ml tubes, No. of holes: 2, Depth: 72,4 mm
Supplier: VWR Collection
Description: Block heater block, VWR®, Aluminium block, For: 0,5 ml tubes, No. of holes: 54
Supplier: VWR Collection
Description: Block heater block, VWR®, Interchangeable tube block, Well size: Ø 7,9 mm, For: 30×0,5 ml tubes**, No. of holes: 30, Depth: 25,7 mm
Supplier: VWR Collection
Description: Block heater block, VWR®, Single block, Well size: Ø 19,7 mm, For: 19 mm vials, No. of holes: 12, Depth: 45,0 mm
Supplier: VWR Collection
Description: Multi-purpose, high wattage, constant temperature, VWR® analogue block heaters are economical, versatile and compact.
Catalog Number: (460-0249)
Supplier: VWR Collection
Description: The combination of heating/cooling and shaking makes the Thermal Shake lite microtube shaking incubator ideal for many life science research ap...
UOM: 1 * 1 items
Supplier: VWR Collection
Description: These mini block heaters are ideal for applications that require accurate, repeatable results. These personal-sized units take up minimal bench space....
Waste Reducing
Catalog Number: (460-3265)
Supplier: VWR Collection
Description: Designed for applications that require repeatable results and superior temperature stability. These multipurpose units are ideal for incubation and ac...
UOM: 1 * 1 items
Supplier: VWR Collection
Description: Solid blocks: For use as a low temperature hotplate or for custom drilling.
Supplier: VWR Collection
Description: The cooling thermal shake touch is designed for applications that require consistent, precise results. With heating, cooling and shaking capabilities,...
Supplier: VWR Collection
Description: Multi-purpose, high wattage, constant temperature, VWR® digital block heaters are economical, versatile and compact. These digital models provide repr...
Waste Reducing
Supplier: VWR Collection
Description: The Thermal Shake Touch is designed for applications that require consistent, precise results. With heating and shaking capabilities, this low profile...
Catalog Number: (460-0353)
Supplier: VWR Collection
Description: Multi-purpose, high wattage, constant temperature, VWR® digital block heater which is economical, versatile and compact. This digital model, with heat...
UOM: 1 * 1 items
Waste Reducing
Catalog Number: (460-3261)
Supplier: VWR Collection
Description: These multipurpose units are ideal for incubation and activation of cultures, enzyme reactions, blood urea nitrogen determinations, immunoassays, melt...
UOM: 1 * 1 items
Waste Reducing
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 80
no targeter for Bottom