You Searched For: Barometers
10  results were found
SearchResultCount:"10"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (620-2630)
Supplier: VWR Collection
Description: The VWR® Traceable® excursion trac USB data logging barometer simultaneously monitors barometric pressure, temperature and humidity.
UOM: 1 * 1 items
Catalog Number: (620-1922)
Supplier: VWR Collection
Description: This compact, lightweight meter can measure barometric pressure, temperature and altitude. It graphically displays the barometric pressure trend, and ...
UOM: 1 * 1 items
Catalog Number: (620-2632)
Supplier: VWR Collection
Description: The VWR® Traceable® memory loc, USB, data logging barometer simultaneously monitors barometric pressure, temperature and humidity. This device can be ...
UOM: 1 * 1 items
Catalog Number: (628-0041)
Supplier: VWR Collection
Description: This barometer with triple-scale provides a sensitive measure of atmospheric pressure. Corrosion-proof, nickel-chrome.
UOM: 1 * 1 items
Catalog Number: (620-2330)
Supplier: VWR Collection
Description: This digital barometer monitors barometric pressure, air temperature, relative humidity also barometric pressure trend and time.
UOM: 1 * 1 items
Catalog Number: (620-3303)
Supplier: Testo
Description: Pocket absolute pressure meter. Very handy, small and easy to use, this barometer permits measurements of barometric air pressure by entering on-site ...
UOM: 1 * 1 items
Catalog Number: (GRSG600407)
Supplier: Greisinger
Description: This handheld meter with integrated absolute pressure sensor is suitable for measurements in air and non corrosive and non ionizing gases and liquids.
UOM: 1 * 1 items
Catalog Number: (THIE3.1509.00.000)
Supplier: Thies Clima
Description: Light grey varnished
UOM: 1 * 1 items
Catalog Number: (765-1090)
Supplier: MÍLLER THERM
Description: Golden scale. Wooden plate Ø: 150 mm.
UOM: 1 * 1 items
Catalog Number: (763-0412)
Supplier: MÍLLER THERM
Description: This baro, hygro and thermometer made from wood is designed for measurements of temperature, relative humidity and pressure.
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 10 of 10
no targeter for Bottom