Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • In Stock (12)
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "Moisture Balances"
https://si.vwr.com/stibo/smallweb/std.lang.all/40/60/18374060.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"17"
  List View Grid View Easy View (new)
Sort by:
Supplier:  VWR Collection
Description:   Rugged compact instruments for moisture determination of raw materials and finished products in the food, cosmetics, dairy, pharmaceutical, chemical a...
Supplier:  VWR Collection
Description:   Reusable sample dishes are essential for use with moisture analysers
Supplier:  VWR Collection
Description:   Moisture balances with large 5'' colour touch screeen display that gives a user friendly access to advanced applications and internal databases. Test ...
Supplier:  SARTORIUS BALANCES
Description:   The MA100 is an accurate, flexible, configurable model with a user friendly keyboard and software prompts to easily access all the modes of operation ...
Supplier:  SARTORIUS BALANCES
Description:   External calibration weight 30 g ±0,3 mg for MA35
Catalog Number: (611-1480)
Supplier:  SARTORIUS BALANCES
Description:   The range of performance features of this moisture analyser is ideal for daily routine use for in-process control.
UOM:  1 * 1 items
Supplier:  SARTORIUS BALANCES
Description:   Easy to use, efficient, compact moisture analysers, introduced in 2014, to give fast repeatable results with an improved user interface.
Supplier:  OHAUS
Description:   Sample pans, reusable
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8  of 17
  1  2  3  Next