Industrial scales, Defender 3000 series
Supplier: Ohaus
Defender 3000
611-5066EA 590 EUR
611-5066 611-2655 611-2654 611-2653 611-4983 611-2652 611-2651 611-2650
Industrial scales, Defender 3000 series
Balances and Scales
The Defender 3000 industrial scales are designed for general purpose weighing requirements and features a stainless steel pan with painted steel frame, IP 67 aluminium load cell and painted steel column support for the ABS indicator. Software applications include parts counting and five weighing units, which makes the scales ideal for shipping and inventory control.
  • Mains or built-in rechargeable battery operation with auto-off
  • Indicator features a backlit LCD display
  • Levelling feet and bubble
  • External calibration