You Searched For: Anemometers
6  results were found
SearchResultCount:"6"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: Testo
Description: The compact 100 mm vane anemometer testo 417 is the ideal solution for simple, fast and precise determination of flow velocity and volume flow at air ...
Supplier: Testo
Description: These Smart Probes are designed to be used with smartphones and tablets. Users can read the measurements using the testo Smart Probes App installed on...
Supplier: Testo
Description: Handy, small and simple, these air velocity meters can measure temperature and wind speed, as well as humidity (620-3308 only). They can also perform ...
Catalog Number: (TEST0563.0425)
Supplier: Testo
Description: The testo 425 digital hot wire anemometer with App connection is fast, easy and precise companion. All relevant calculations, such as volume flow or ...
UOM: 1 * 1 items
Catalog Number: (620-3311)
Supplier: Testo
Description: The 405 is a pocket anemometer with telescopic handle to facilitate the measurement of air flow, volume flow and temperature.
UOM: 1 * 1 items
Catalog Number: (TEST0563.0416)
Supplier: Testo
Description: The compact 16 mm vane anemometer testo 416 with telescopic probe and App connection, we can carry out flow measurements in the ventilation ducts of a...
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 6 of 6
no targeter for Bottom