Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • In Stock (18)
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
Flask Weights
https://si.vwr.com/stibo/smallweb/std.lang.all/52/44/18945244.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"9"
  List View Easy View (new)
Sort by:
Supplier:  BOCHEM
Description:   PVC covered steel rings.
Supplier:  HEATHROW
Description:   Lead core, with blue vinyl coating. Open or closed design. Open design is also suitable for vessels with lateral connections. For stabilising glass or...
Supplier:  KLEINFELD GMBH & CO.
Description:   Lead with PVC coating. These cuffs are flexible. They can be opened, closed or adapted to the size of the bottle.
Supplier:  Bel-Art
Description:   Soft lead sheet coated with flexible Vikem® vinyl has many uses for stabilizing laboratory equipment in water baths or any application where extra wei...
Catalog Number: (214-1917)
Supplier:  KOEHLER TECHNISCHE PRODUKTEN
Description:   Weight, Flask Immersion, Lead with red PVC coating, Open C-shaped ring, For: Flasks 750 - 2000 ml, 1190 g, Ø int.: 100 mm
UOM:  1 * 1 items
Supplier:  Bel-Art
Description:   Lead core, with green vinyl coating. Open design (C shaped) is suitable for vessels with lateral connections. For stabilising glass or plastic vessels...
Supplier:  DELTALAB
Description:   Weight, Flask Immersion, vinyl coating, Closed ring, For: Flasks 500 - 2000 ml, Ø int.: 57 mm
Supplier:  Bel-Art
Description:   Lead core, with Vikem® vinyl coating.
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8  of 9
  1  2  Next 
A new look and a renewed commitment
Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.
Learn More Start Shopping