You Searched For: Capillary Viscometers
16  results were found
SearchResultCount:"16"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: SI Analytics
Description: Ostwald viscometers suitable for measurements of transparent liquids manual measurements, foaming liquids and liquid mixture with highly volatile comp...
Supplier: SI Analytics
Description: Cannon-Fenske viscometers for automatic measurements especially in routine analyses.
Supplier: SI Analytics
Description: Viscometers, with suspended ball level, for determination of relative or absolute kinematic viscosity.
Supplier: SI Analytics
Description: These Micro-Ostwald viscometers are available as calibrated for either manual or automatic measurements and uncalibrated models.
Supplier: SI Analytics
Description: Ubbelohde viscometers for manual measurements of transparent liquids.
Supplier: SI Analytics
Description: Ubbelohde viscometers with suspended ball level used to determine the limit viscosity of polymers.
Supplier: SI Analytics
Description: Ubbelohde viscometers for the determination of absolute and relative viscosity of liquids with Newtonian flow behaviour.
Supplier: SI Analytics
Description: Viscometers with suspended ball level for determination of relative or absolute kinematic viscosity.
Supplier: SI Analytics
Description: Calibrated for automatic measurement, viscometers with suspended ball level for determination of absolute and relative viscosity of liquids with Newto...
Supplier: SI Analytics
Description: Viscometers with suspended ball level for determination of relative viscosity of liquids with Newtonian flow behaviour.
Supplier: SI Analytics
Description: Reverse flow viscometers calibrated with constant.
Supplier: SI Analytics
Description: Capillary viscometers calibrated for automatic measurements in routine applications.
Supplier: SI Analytics
Description: Cannon-Fenske viscometers for manual measurements especially in routine analyses.
Supplier: SI Analytics
Description: Viscometers with suspended ball level for determination of relative viscosity of liquids with Newtonian flow behaviour.
Supplier: SI Analytics
Description: Ubbelohde viscometers with suspended ball level for determination of relative or absolute kinematic viscosity.
Supplier: DWK Life Sciences
Description: This viscometer requires calibration against a standard liquid of known viscosity or against a second viscometer with known constant.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 16
no targeter for Bottom