You Searched For: Valves
179  results were found
SearchResultCount:"179"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: Avantor Fluid Handling
Description: Build leakproof fluid handling systems.
Supplier: Avantor Fluid Handling
Description: Regulate flow without any leaking.
Supplier: Avantor Fluid Handling
Description: Regulate flow without any leaking.
Sale
Supplier: Avantor Fluid Handling
Description: Regulate flow without any leaking.
Supplier: Avantor Fluid Handling
Description: Enhance fluid flow with a larger bore than standard luer stopcocks.
Catalog Number: (MFLX30526-34)
Supplier: Avantor Fluid Handling
Description: Pressure rating up to 1050 psi.
UOM: 1 * 10 items
Supplier: Avantor Fluid Handling
Description: Regulate flow without any leaking.
Catalog Number: (MFLX30526-34BLK)
Supplier: Avantor Fluid Handling
Description: Pressure rating up to 1050 psi.
UOM: 1 * 200 items
Catalog Number: (HICHHI-128)
Supplier: Avantor
Description: Avantor® Hichrom, Back pressure regulator, Pressure: 5,17 bar
UOM: 1 * 1 items
Supplier: ENTEGRIS INC
Description: This 3-way valve (patent-pending) has an all-PFA valve body and high purity fluoropolymer wetted components providing a noncontaminating fluid stream ...
Supplier: ENTEGRIS INC
Description: These components are constructed of high purity PFA and PTFE materials for wetted parts and PVDF, PEEK™ and PTFE u-cup with Viton® energizer for non w...
Supplier: ENTEGRIS INC
Description: O-ring free design, ¹/₂ or ¹/₄" orifice.
Supplier: ENTEGRIS INC
Description: Two- or three-way diaphragm valves, ¹/₄" orifice, normally closed.
Supplier: ENTEGRIS INC
Description: Available compliant to SEMI Standard F57-0312, the specification for polymer components used in ultrapure water and liquid chemical distribution syste...
Supplier: ENTEGRIS INC
Description: Available compliant to SEMI Standard F57-0312, the specification for polymer components used in ultrapure water and liquid chemical distribution syste...
Supplier: ENTEGRIS INC
Description: Available compliant to SEMI Standard F57-0312, the specification for polymer components used in ultrapure water and liquid chemical distribution syste...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 179
no targeter for Bottom