You Searched For: NMR Tubes
28  results were found
SearchResultCount:"28"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (WILM522-PP-7-5)
Supplier: SP Industries
Description: Borosilicate glass, Class A, ASTM Type 1.
UOM: 1 * 5 items
Catalog Number: (WILMWG-3000-7-50)
Supplier: SP Industries
Description: High-throughput NMR tubes are made of borosilicate glass Type I, class A.
UOM: 1 * 50 items
Catalog Number: (WILMWG-221T)
Supplier: SP Industries
Description: Q-band is the most popular frequency for pulsed and cw-EPR after X-band.
UOM: 1 * 10 items
Supplier: SP Industries
Description: Quartz (natural quartz).
Supplier: SP Industries
Description: Quartz (natural quartz).
Supplier: SP Industries
Description: Borosilicate glass, class A, ASTM Type 1.
Supplier: SP Industries
Description: Borosilicate glass, Class A, ASTM Type 1.
Supplier: SP Industries
Description: Borosilicate glass, Class A, ASTM Type 1.
Supplier: SP Industries
Description: NMR tubes, economy, 5 mm, borosilicate glass ASTM E438, Type I, class B, with disposable cap.
Supplier: SP Industries
Description: These Wilmad-LabGlass NMR tubes are supplied with authentic closed Bruker® SampleJet® caps without code.
Supplier: SP Industries
Description: Borosilicate glass, Class A, ASTM Type 1.
Supplier: SP Industries
Description: Pyrex® glass, Class A, ASTM type 1.
Supplier: SP Industries
Description: Clear Fused Quartz (CFQ).
Supplier: SP Industries
Description: Borosilicate glass
Supplier: SP Industries
Description: Borosilicate glass, class B, ASTM E438 Type 1
Supplier: SP Industries
Description: Double the writing area allows for easy sample marking and identification.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 28
no targeter for Bottom