You Searched For: Microtubes
16  results were found
SearchResultCount:"16"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: Avantor
Description: Safe sample storage system.
Sale
Catalog Number: (KIMA18167)
Supplier: KIMA SAS DI CHIARIN RENZO & C
Description: PP, translucent.
UOM: 1 * 2.000 items
Supplier: Corning
Description: PP, 100% virgin. The Axygen 96-well mini tube system is designed to work with 8-strip or 12-strip plug caps.
Supplier: Corning
Description: PP. The 1D/2D storage line is an innovative product with both a linear 1D and 2D barcodes etched on each tube.
Supplier: Thermo Fisher Scientific
Description: Microtube racke system consists of 1,1 ml-tall microtubes supplied pre-racked in boxes to match a standard 96-well microplate footprint.
Supplier: Brand
Description: PP tubes and PE caps, non-sterile.
Supplier: Simport Scientific
Description: These racks contain 96 removable PP tubes, either placed individually or in a 8×12 configuration.
Supplier: Simport Scientific
Description: These racks contain 96 removable PP square tubes in a 8×12 configuration, each having a 2,1 ml capacity (2 ml when capped). Tubes are also available s...
Supplier: Thermo Fisher Scientific
Description: Tubes made of PP, natual, caps made of PE, natural.
Supplier: Simport Scientific
Description: PP tube and rack system with tubes held in a standard 8×12 configuration.
Supplier: RATIOLAB
Description: Translucent polypropylene (PP) with very low protein binding.
Sale
Supplier: RATIOLAB
Description: Plastic rack with cardboard box.
Catalog Number: (SIMPT110-15)
Supplier: Simport Scientific
Description: Made of PP, translucent.
UOM: 1 * 120 items
Supplier: KARTELL
Description: Ideal for samples analysis.
Catalog Number: (391-9030)
Supplier: Bel-Art Products, a Part of SP
Description: Made of virgin, medical grade polypropylene.
UOM: 1 * 4.800 items
Supplier: SARSTEDT
Description: PP, translucent
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 16
no targeter for Bottom